Learn More
Medchemexpress LLC Berubicin (hydrochloride) | 293736-67-1 | 95.08% | 670.10 | 5 MG
SDP
Catalog No. 5000360252
Encompass_Preferred
Berubicin hydrochloride is a Doxorubicin analog capable of crossing the blood-brain barrier. It inhibits P-gp and MRP1-mediated efflux, suppressing glioblastoma multiforme (GBM). This compound exhibits toxic effects on leukemia cells by activating nuclear factor κB (NF-κB) and induces apoptosis in neuroblastoma cells. It is suitable for studying nervous system-related tumors.
  • Can cross the blood-brain barrier
  • Inhibits P-gp and MRP1-mediated efflux
  • Suppresses glioblastoma multiforme (GBM)
  • Exerts toxic effects on leukemia cells by activating nuclear factor κB (NF-κB)
  • Induces apoptosis in neuroblastoma cells
  • Useful in the study of nervous system-related tumors
Catalog No. 50-003-60252 Supplier Medchemexpress LLC Supplier No. HY14942A5MG
Berubicin hydrochloride is a Doxorubicin analog capable of crossing the blood-brain barrier. It inhibits P-gp and MRP1-mediated efflux, suppressing glioblastoma multiforme (GBM). This compound exhibits toxic effects on leukemia cells by activating nuclear factor κB (NF-κB) and induces apoptosis in neuroblastoma cells. It is suitable for studying nervous system-related tumors.
  • Can cross the blood-brain barrier
  • Inhibits P-gp and MRP1-mediated efflux
  • Suppresses glioblastoma multiforme (GBM)
  • Exerts toxic effects on leukemia cells by activating nuclear factor κB (NF-κB)
  • Induces apoptosis in neuroblastoma cells
  • Useful in the study of nervous system-related tumors