Learn More
Medchemexpress LLC Berubicin hydrochloride | 293736-67-1 | 95.1% | 670.10 | 1 MG
SDP
Catalog No. 5000360835
Encompass_Preferred
Berubicin hydrochloride is an analog of Doxorubicin that can cross the blood-brain barrier. It inhibits P-gp and MRP1-mediated efflux and suppresses glioblastoma multiforme (GBM). This product exerts toxic effects on leukemia cells by activating nuclear factor κB (NF-κB) and induces apoptosis in neuroblastoma cells. It can be used in the study of tumors related to the nervous system.
  • Crosses the blood-brain barrier
  • Inhibits P-gp and MRP1-mediated efflux
  • Suppresses glioblastoma multiforme (GBM)
  • Activates nuclear factor κB (NF-κB)
  • Induces apoptosis in neuroblastoma cells
  • More potent activity than Doxorubicin in activating NF-κB and inhibiting DNA synthesis
  • Prolongs survival time in intracranial orthotopic glioma models in mice
Catalog No. 50-003-60835 Supplier Medchemexpress LLC Supplier No. HY14942A1MG
Berubicin hydrochloride is an analog of Doxorubicin that can cross the blood-brain barrier. It inhibits P-gp and MRP1-mediated efflux and suppresses glioblastoma multiforme (GBM). This product exerts toxic effects on leukemia cells by activating nuclear factor κB (NF-κB) and induces apoptosis in neuroblastoma cells. It can be used in the study of tumors related to the nervous system.
  • Crosses the blood-brain barrier
  • Inhibits P-gp and MRP1-mediated efflux
  • Suppresses glioblastoma multiforme (GBM)
  • Activates nuclear factor κB (NF-κB)
  • Induces apoptosis in neuroblastoma cells
  • More potent activity than Doxorubicin in activating NF-κB and inhibiting DNA synthesis
  • Prolongs survival time in intracranial orthotopic glioma models in mice