Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium, AAS standard solution, Specpure™ Be 1000μg/mL

Catalog No. p-4408531
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88053AE 100 mL
AA88053AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88053AE Supplier Thermo Scientific Chemicals Supplier No. 088053AE
Only null left
Add to Cart

CAS: 19049-40-2 | Be in 5% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19049-40-2 , 7732-18-5
Molecular Formula Be in 5% HNO3
MDL Number MFCD00134032

Specifications

Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Quantity 100 mL
Grade AAS Standard
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Beryllium oxide acetate: 1.13%; Water: 93.87%
Chemical Name or Material Beryllium
Hazard Category H290-H314-H317-H332-H335-H350i-H373
Precautionary Statement P201-P202-P234-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.