Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium, plasma standard solution, Specpure™ Be 1000μg/mL

Catalog No. AA13848AD
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13848AD 50 mL
AA13848AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA13848AD Supplier Thermo Scientific Chemicals Supplier No. 013848AD
Add to Cart

missing translation for 'validMainframeMsg'

Be4O(C2H3O2)6 in 5% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134032

Specifications

Physical Form Liquid
Molecular Formula Be4O(C2H3O2)6 in 5% HNO3
Concentration Be 1000 μg/mL
UN Number UN3264
Type Analytical Standard
Quantity 50 mL
Grade Specpure
Chemical Name or Material Beryllium, plasma standard solution, Specpure™
Hazard Category H290-H314-H317-H332-H335-H350i-H373
Hazard Statement GHS H StatementH350-H373-H314-H317;May cause cancer.May cause damage to organs through prolonged or repeated exposure.Causes severe skin burns and eye damage.May cause an allergic skin reaction.
Precautionary Statement P201-P202-P234-P260-P264b-P271-P272-P280g-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.