Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllium Thinfoil, 0.005mm (0.0002 in.) thick, 99.8% (metals basis), Thermo Scientific™

Catalog No. AA415550T
Change view
Click to view available options
12.5x12.5mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA415550T 12.5x12.5mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA415550T Supplier Thermo Scientific Chemicals Supplier No. 415550T

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-41-7
Molecular Formula Be
MDL Number MFCD00134032
Quantity 12.5 x 12.5mm
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
SMILES [Be]
IUPAC Name beryllium
Molecular Weight (g/mol) 9.012
PubChem CID 5460467
ChEBI CHEBI:33783
Percent Purity 99.8%
Odor Odorless
Weight ≈0.0015g/12.5x12.5mm
Assay (metals basis)
Chemical Name or Material Beryllium Thinfoil, 0.005mm (0.0002 in.) thick, Not certified pinhole free
Show More Show Less
Hazard Category H301-H315-H317-H319-H330-H335-H350i-H372
Hazard Statement GHS H Statement
H301-H330-H350-H372-H315-H319-H317-H335
Toxic if swallowed.
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
EINECSNumber 231-150-7
RTECSNumber DS1750000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.