Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beryllon II, TCI America™
SDP

Catalog No. B04521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B04521G Supplier TCI America Supplier No. B04521G

Chemical Identifiers

CAS 51550-25-5
Molecular Formula C20H10N2Na4O15S4
Molecular Weight (g/mol) 738.50
MDL Number MFCD00046409
InChI Key CBWXTVSARCWSMJ-QNKYWQQSSA-J
Synonym beryllon, 2,7-naphthalenedisulfonic acid, 4,5-dihydroxy-3-2-8-hydroxy-3,6-disulfo-1-naphthalenyl diazenyl-, sodium salt 1:4, tetrasodium 4,5-dihydroxy-3-e-2-8-hydroxy-3,6-disulfonatonaphthalen-1-yl diazen-1-yl naphthalene-2,7-disulfonate
PubChem CID 44135400
IUPAC Name tetrasodium 4-hydroxy-5-{2-[(2E)-8-hydroxy-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].OC1=CC(=CC2=CC(=CC(N\N=C3/C(=O)C4=C(C=C(C=C4O)S([O-])(=O)=O)C=C3S([O-])(=O)=O)=C12)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 738.50
Physical Form Crystalline Powder
Chemical Name or Material Beryllon II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.