Learn More
Selleck Chemical LLC Berzosertib
SDP
Catalog No. 5000004842
Encompass_Preferred
Berzosertib (VE-822 VX970 M6620) is an ATR inhibitor with IC50 of 19 nM in HT29 cells
Catalog No. 50-000-04842 Supplier Selleck Chemical LLC Supplier No. S7102200MG
Berzosertib (VE-822 VX970 M6620) is an ATR inhibitor with IC50 of 19 nM in HT29 cells