Learn More
Selleck Chemical LLC Berzosertib VE-822-10mg
SDP
Supplier:  Selleck Chemical LLC S710210MG
Encompass_Preferred
Berzosertib (VE-822, VX970, M6620) is an ATR inhibitor with IC50 of 19 nM in HT29 cells.
Catalog No. 50-238-9826