Learn More
Bioworld BES Buffer, 0.5M, pH 7.5, 250 mL
Catalog No. 50255429
Encompass_Preferred
Product DescriptionBES (2-(Bis(2-hydroxyethyl) amino) ethanesulfonic acid) buffer has a concentration of 0.5M and a pH value of 7.5. BES is a zwitterionic buffer used in biochemistry and molecular biology, and has been used to study versatile catalyst precursors for mild hydrocarboxylation of alkanes to carboxylic acids. BES was used to determine that saline buffers minimize the action of interfering factors in the bacterial endotoxins test.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-429 Supplier Bioworld Supplier No. 402200052
Product DescriptionBES (2-(Bis(2-hydroxyethyl) amino) ethanesulfonic acid) buffer has a concentration of 0.5M and a pH value of 7.5. BES is a zwitterionic buffer used in biochemistry and molecular biology, and has been used to study versatile catalyst precursors for mild hydrocarboxylation of alkanes to carboxylic acids. BES was used to determine that saline buffers minimize the action of interfering factors in the bacterial endotoxins test.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.