Learn More
Research Products International Corp BES [N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid], 250 Grams
SDP
Catalog No. 50213170
Encompass_Preferred
For use in preparing electrophoresis buffers. pKa (25° C) 7.1. The working pH Range in aqueous solution is 6.4-7.8
Catalog No. 50-213-170 Supplier Research Products International Corp Supplier No. B32070250.0
For use in preparing electrophoresis buffers. pKa (25° C) 7.1. The working pH Range in aqueous solution is 6.4-7.8