Learn More
Research Products International Corp BES 500 G
SDP
Catalog No. 502230513
Encompass_Preferred
For use in preparing electrophoresis buffers. pKa (25° C) 7.1. The working pH Range in aqueous solution is 6.4-7.8
Catalog No. 50-223-0513 Supplier Research Products International Corp Supplier No. B32070500.0
For use in preparing electrophoresis buffers. pKa (25° C) 7.1. The working pH Range in aqueous solution is 6.4-7.8