Learn More
Sigma Aldrich Fine Chemicals Biosciences BES 99.0 titration250G
Catalog No. 502903274
Encompass
BES (N N-bis(2-hydroxyethyl)-2-aminoethanesulfonic acid) is a useful secondary standard biochemical buffer. Useful pH range for BES is 6.4 to 7.8. It is useful for diagnostic assay manufacturing industry. BES a sulfonic acid-containing cross-linking agent induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes.
Catalog No. 50-290-3274 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B9879250G
BES (N N-bis(2-hydroxyethyl)-2-aminoethanesulfonic acid) is a useful secondary standard biochemical buffer. Useful pH range for BES is 6.4 to 7.8. It is useful for diagnostic assay manufacturing industry. BES a sulfonic acid-containing cross-linking agent induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes.