Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BES, 99+%, MP Biomedicals™

Catalog No. ICN10092705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10092705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10092705 Supplier MP Biomedicals, Inc Supplier No. 0210092705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym n,n-bis 2-hydroxyethyl-2-aminoethanesulfonic acid, bes, ethanesulfonic acid, 2-bis 2-hydroxyethyl amino, 2-bis 2-hydroxyethyl amino ethanesulfonic acid, n,n-bis 2-hydroxyethyl taurine, n,n-bis hydroxyethyl-2-aminoethanesulfonic acid, bes buffering agent, taurine, n,n-bis 2-hydroxyethyl, 2-bis 2-hydroxyethyl amino-ethanesulfonicacid
PubChem CID 73243
ChEBI CHEBI:39045
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99+%
Formula Weight 213.3
Chemical Name or Material BES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.