Learn More
Alkali Scientific BES [N,N bis(2 Hydroxyethyl) 2 aminoethanesulfonic acid], 500 Grams
SDP
Catalog No. 5000661161
Encompass_Preferred
BES (N,N bis(2 Hydroxyethyl) 2 aminoethanesulfonic acid) is a biochemical reagent used as a pH buffer in research and laboratory applications. This 500-gram volume is ideal for the preparation of buffer solutions with stable pH levels, particularly for biochemical assays and protein analysis. BES is used widely in enzyme studies, especially where precise pH control is required. It is valued for its ability to maintain a consistent pH in solutions that mimic physiological environments. The reagent’s high buffering capacity ensures reliable performance in a variety of applications, including cell culture and chromatography processes, where pH balance is crucial for accurate results.
Catalog No. 50-006-61161 Supplier Alkali Scientific Supplier No. BEA004CP
BES (N,N bis(2 Hydroxyethyl) 2 aminoethanesulfonic acid) is a biochemical reagent used as a pH buffer in research and laboratory applications. This 500-gram volume is ideal for the preparation of buffer solutions with stable pH levels, particularly for biochemical assays and protein analysis. BES is used widely in enzyme studies, especially where precise pH control is required. It is valued for its ability to maintain a consistent pH in solutions that mimic physiological environments. The reagent’s high buffering capacity ensures reliable performance in a variety of applications, including cell culture and chromatography processes, where pH balance is crucial for accurate results.