Learn More
Bioworld BES, 1M, pH 7.0, 200 mL
Catalog No. 50255388
Encompass_Preferred
BES bufferAlso known as N,N-Bis(hydroxyethyl)-2aminoethanesulfonic acid.Technical Information:pKA (25 C): 7.1Heavy Metals: <0.0005%. Appearance: LiquidBoiling Point : 404.34 °C (Predicted)Density : 1.44 g/cm3 (Predicted)Melting Point: 152-154 °CPH Value: 6.4-7.8
Catalog No. 50-255-388 Supplier Bioworld Supplier No. 402000282
BES bufferAlso known as N,N-Bis(hydroxyethyl)-2aminoethanesulfonic acid.Technical Information:pKA (25 C): 7.1Heavy Metals: <0.0005%. Appearance: LiquidBoiling Point : 404.34 °C (Predicted)Density : 1.44 g/cm3 (Predicted)Melting Point: 152-154 °CPH Value: 6.4-7.8