Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BES sodium | 66992-27-6 | 97.0% | 25 G
SDP

Catalog No. 5000414095
Encompass_Preferred

BES sodium is a secondary standard biochemical buffer with an effective pH range of 6.4 to 7.8 (pKa: 7.15 at 20 °C). It is used in the diagnostic testing area.

  • Biochemical buffer
  • Effective pH range: 6.4 to 7.8 (pKa: 7.15 at 20 °C)
  • Used in diagnostic testing
  • Appearance: Solid
  • Color: White to off-white
  • Target: Biochemical assay reagents
  • Pathway: Others

Catalog No. 50-004-14095 Supplier Medchemexpress LLC Supplier No. 25GHYD0867A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BES sodium is a secondary standard biochemical buffer with an effective pH range of 6.4 to 7.8 (pKa: 7.15 at 20 °C). It is used in the diagnostic testing area.

  • Biochemical buffer
  • Effective pH range: 6.4 to 7.8 (pKa: 7.15 at 20 °C)
  • Used in diagnostic testing
  • Appearance: Solid
  • Color: White to off-white
  • Target: Biochemical assay reagents
  • Pathway: Others

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.