Learn More
Medchemexpress LLC Besifloxacin (Hydrochloride) | 405165-61-9 | 98.5% | 430.30 | 25 MG
SDP
Catalog No. 5000385127
Encompass_Preferred
Besifloxacin Hydrochloride is a fourth-generation fluoroquinolone antibiotic that functions as a DNA gyrase and topoisomerase IV inhibitor. It exhibits broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains and helps reduce the incidence of drug resistance. It also possesses anti-inflammatory properties and can be utilized in bacterial conjunctivitis research.
  • Functions as a DNA gyrase and topoisomerase IV inhibitor.
  • Exhibits broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains.
  • Reduces the incidence of drug resistance.
  • Possesses anti-inflammatory activity.
  • Can be used in bacterial conjunctivitis research.
Catalog No. 50-003-85127 Supplier Medchemexpress LLC Supplier No. 25MGHY17028
Besifloxacin Hydrochloride is a fourth-generation fluoroquinolone antibiotic that functions as a DNA gyrase and topoisomerase IV inhibitor. It exhibits broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains and helps reduce the incidence of drug resistance. It also possesses anti-inflammatory properties and can be utilized in bacterial conjunctivitis research.
  • Functions as a DNA gyrase and topoisomerase IV inhibitor.
  • Exhibits broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains.
  • Reduces the incidence of drug resistance.
  • Possesses anti-inflammatory activity.
  • Can be used in bacterial conjunctivitis research.