Learn More
Medchemexpress LLC Besifloxacin hydrochloride | 405165-61-9 | 98.5% | 430.30 | 5 MG
SDP
Catalog No. 5000384296
Encompass_Preferred
Besifloxacin Hydrochloride is a fourth generation fluoroquinolone antibiotic. It functions as a DNA gyrase and topoisomerase IV inhibitor. This compound demonstrates broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains, and it helps reduce the incidence of drug resistance. Additionally, Besifloxacin Hydrochloride exhibits anti-inflammatory activity and is suitable for bacterial conjunctivitis research.
  • Fourth generation fluoroquinolone antibiotic.
  • DNA gyrase and topoisomerase IV inhibitor.
  • Broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains.
  • Reduces the incidence of drug resistance.
  • Anti-inflammatory activity.
  • Used in bacterial conjunctivitis research.
Catalog No. 50-003-84296 Supplier Medchemexpress LLC Supplier No. HY170285MG
Besifloxacin Hydrochloride is a fourth generation fluoroquinolone antibiotic. It functions as a DNA gyrase and topoisomerase IV inhibitor. This compound demonstrates broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains, and it helps reduce the incidence of drug resistance. Additionally, Besifloxacin Hydrochloride exhibits anti-inflammatory activity and is suitable for bacterial conjunctivitis research.
  • Fourth generation fluoroquinolone antibiotic.
  • DNA gyrase and topoisomerase IV inhibitor.
  • Broad-spectrum antibacterial activity against Gram-negative and Gram-positive aerobic and anaerobic strains.
  • Reduces the incidence of drug resistance.
  • Anti-inflammatory activity.
  • Used in bacterial conjunctivitis research.