Learn More
Medchemexpress LLC HY-19447 5mg Medchemexpress, Besifovir CAS:441785-25-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY194475MG
Encompass_Preferred
Medchemexpress, HY-19447 5mg Besifovir CAS:441785-25-7 Besifovir (LB80331), a parent drug converted by LB80380, further metabolizes to its active form, LB80317. LB80380 is potent antiviral agent against hepatitis B virus (HBV) [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2832