Learn More
MSC Betco Liquid Abrasive Creme Cleanser, 32 oz, Mint
Catalog No. 502839102
Encompass
This liquid creme cleanser quickly removes embedded rust, soap scum, stains, grease, oxidation, hard water minerals, and filth from a variety of surfaces. It contains jeweler grade abrasive that will not scratch surfaces like scouring powders, making it suitable for delicate applications.
  • Thick clinging formula for vertical surfaces.
  • Penetrates, scours, brightens, and polishes without heavy agitation.
  • Free rinsing.
  • Fresh mint fragrance.
  • Will not scratch surfaces like scouring powders.
  • Contains bleaching compounds, detergents, and wetting agents.
Catalog No. 50-283-9102 Supplier MSC Supplier No. 72969785
This liquid creme cleanser quickly removes embedded rust, soap scum, stains, grease, oxidation, hard water minerals, and filth from a variety of surfaces. It contains jeweler grade abrasive that will not scratch surfaces like scouring powders, making it suitable for delicate applications.
  • Thick clinging formula for vertical surfaces.
  • Penetrates, scours, brightens, and polishes without heavy agitation.
  • Free rinsing.
  • Fresh mint fragrance.
  • Will not scratch surfaces like scouring powders.
  • Contains bleaching compounds, detergents, and wetting agents.