Learn More
Enterprise Technology Solutions Best Manufacturing Company Safety Gloves, Natural Rubber, Medium, Blue/Gray
SDP
Catalog No. 5000828548
Encompass_Preferred
Palm-dipped, blue-colored natural rubber gloves provide durable protection without sacrificing long-wearing comfort. Cotton/polyester lining provides extremely comfortable fit. The rough textured lightweight natural rubber coating provides outstanding grip. An elastic knit-wrist secures the glove above the wrist for comfort and preventing debris from entering the glove. Knuckles are not coated to provide dexterity and coolness.
Catalog No. 50-008-28548 Supplier Enterprise Technology Solutions Supplier No. BSM30008
Palm-dipped, blue-colored natural rubber gloves provide durable protection without sacrificing long-wearing comfort. Cotton/polyester lining provides extremely comfortable fit. The rough textured lightweight natural rubber coating provides outstanding grip. An elastic knit-wrist secures the glove above the wrist for comfort and preventing debris from entering the glove. Knuckles are not coated to provide dexterity and coolness.