Learn More
Electron Microscopy Sciences Best's Carmine Stain Solution 250 ML
SDP
Catalog No. 501901633
Encompass_Preferred
Bests Carmine Stain solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used for selectively staining glycogen. (Keep Refrigerated)
Catalog No. 50-190-1633 Supplier Electron Microscopy Sciences Supplier No. 2602502
Bests Carmine Stain solution is a prepared, ready-to-use, high quality staining solution that saves time and effort and produces consistent uniform results. It is used for selectively staining glycogen. (Keep Refrigerated)