Learn More
MSC TESA Brown and Sharpe BesTest Horizontal Dial Indicator, 0.0200 in. Range, 0-10-0 Reading, 0.000500 in. Graduation, 1 ea.
Catalog No. 5000515322
Encompass
The BesTest horizontal dial test indicator is a high-precision dimensional measuring tool designed for accurate surface testing and alignment tasks. Built with a robust monobloc housing and a jeweled movement utilizing 7 rubies, it ensures reliable performance in demanding environments. This indicator features bi-directional measuring capabilities with automatic reversal and maintains a continuous clockwise pointer rotation for clear, error-free readings.
  • Lever type design with integrated shockproof system
  • Ball-bearing lever system allowing the measuring insert to swivel through 240°
  • Insensitive to magnetic fields and corrosion resistant
  • Monobloc construction for exceptional durability
  • Very low measuring force
  • White dial face with 0 to 10 to 0 reading
  • Includes plastic case, mounting rod, wrench, and three 0.08 in. inserts
Catalog No. 50-005-15322 Supplier MSC Supplier No. 97437305
The BesTest horizontal dial test indicator is a high-precision dimensional measuring tool designed for accurate surface testing and alignment tasks. Built with a robust monobloc housing and a jeweled movement utilizing 7 rubies, it ensures reliable performance in demanding environments. This indicator features bi-directional measuring capabilities with automatic reversal and maintains a continuous clockwise pointer rotation for clear, error-free readings.
  • Lever type design with integrated shockproof system
  • Ball-bearing lever system allowing the measuring insert to swivel through 240°
  • Insensitive to magnetic fields and corrosion resistant
  • Monobloc construction for exceptional durability
  • Very low measuring force
  • White dial face with 0 to 10 to 0 reading
  • Includes plastic case, mounting rod, wrench, and three 0.08 in. inserts