Learn More
BEST SANITIZERS Best Refillable Wiping System, 90 ct Heavy Duty
Catalog No. NC0555030
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers refillable wiping system comes with 6 canisters to a case. Wipes are available as a 90 count canister (extra heavy duty wipes).When the canister goes empty, just reorder the 6-pack of refill rolls and reuse the durable canister to save money and reduce waste. The wipes in each canister come dry. Add 1.25 quarts of either Alpet D2, Alpet D2 Quat-Free, Alpet E3 Plus or the product of your choice to the wipes to create the wiping system of your choosing.
This item is not returnable. View return policy
Catalog No. NC0555030 Supplier BEST SANITIZERS Supplier No. SS10005P
This item has a minimum qty of 2 per supplier requirements.
Best Sanitizers refillable wiping system comes with 6 canisters to a case. Wipes are available as a 90 count canister (extra heavy duty wipes).When the canister goes empty, just reorder the 6-pack of refill rolls and reuse the durable canister to save money and reduce waste. The wipes in each canister come dry. Add 1.25 quarts of either Alpet D2, Alpet D2 Quat-Free, Alpet E3 Plus or the product of your choice to the wipes to create the wiping system of your choosing.