Learn More
Aviva Systems Biology Corp BEST3 Antibody - N-terminal region (ARP50109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BEST3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BEST3. Alternate names: MGC13168, MGC40411, VMD2L3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60260