Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestatin, 100%, MP Biomedicals™

Catalog No. ICN15284401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15284401 1 mg
ICN15284410 10 mg
ICN15284425 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15284401 Supplier MP Biomedicals, Inc Supplier No. 0215284401
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58970-76-6
Molecular Formula C16H24N2O4
Molecular Weight (g/mol) 308.38
MDL Number MFCD00083262
InChI Key VGGGPCQERPFHOB-MCIONIFRSA-N
Synonym bestatin, ubenimex, ubenimexum latin, ubenimex inn:jan, bestatin;ubenimex, s-2-2s,3r-3-amino-2-hydroxy-4-phenylbutanamido-4-methylpentanoic acid, ubenimex bestatin, chembl29292, 2s,3r-3-amino-2-hydroxy-4-phenylbutanoyl-l-leucine, 3-r-amino-2-s-hydroxy-4-phenylbutanoyl-s-leucine
PubChem CID 72172
IUPAC Name (2R)-2-[(2S,3R)-3-amino-2-hydroxy-4-phenylbutanamido]-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(=O)[C@@H](O)[C@H](N)CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Assay Percent Range 1
Quantity 1 mg
Formula Weight 308.4
Physical Form Powder
Chemical Name or Material Bestatin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.