Learn More
Apexbio Technology LLC Bestatin, 10mg. Cas: 58970-76-6 MFCD: MFCD00083262. Aminopeptidase inhibitor
SDP
Supplier:  Apexbio Technology LLC A257510
Encompass_Preferred
Ubenimex(Bestatin) is a specific inhibitor of aminopeptidase B and leucine aminopeptidase. It did not show any inhibition of aminopeptidase A, trypsin, chymotrypsin, elastase, papain, pepsin or thermolysin. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4747