Learn More
Bioworld Bestatin (Ubenimex), 10 mg
Catalog No. 50198750
Encompass_Preferred
Bestatin (Ubenimex) is a competitive protease inhibitor - a specific inhibitor of aminopeptidase B and leucine aminopeptidase. Alternative name(s)/Synonyms: Ubenimex Chemical Name: N-[(2S,3R)-3-Amino-2-hydroxy-1-oxo-4-phenylbutyl]-L-leucine Biological Activity: Aminopeptidase inhibitor (Ki = 0.001-90 mM); inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.
Catalog No. 50-198-750 Supplier Bioworld Supplier No. 215200031
Add to Cart
Add to Cart
Bestatin (Ubenimex) is a competitive protease inhibitor - a specific inhibitor of aminopeptidase B and leucine aminopeptidase. Alternative name(s)/Synonyms: Ubenimex Chemical Name: N-[(2S,3R)-3-Amino-2-hydroxy-1-oxo-4-phenylbutyl]-L-leucine Biological Activity: Aminopeptidase inhibitor (Ki = 0.001-90 mM); inhibits enkephalin metabolism and leukotriene A4 hydrolase. Inhibits tumor cell proliferation.