Learn More
Medchemexpress LLC HY-B0134 100mg Medchemexpress, Bestatin Ubenimex CAS:58970-76-6 Purity:98%
SDP
Catalog No. 501639198
Encompass_Preferred
Medchemexpress,HY-B0134 100mg Bestatin Ubenimex CAS:58970-76-6 Formula:C16H24N2O4 Bestatin is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer treatment. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-9198 Supplier Medchemexpress LLC Supplier No. HYB0134100MG
Medchemexpress,HY-B0134 100mg Bestatin Ubenimex CAS:58970-76-6 Formula:C16H24N2O4 Bestatin is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer treatment. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.