Learn More
Medchemexpress LLC Bestatin-d7 (hydrochloride) | 97.7% | 1 MG
SDP
Catalog No. 5000450884
Encompass_Preferred
Bestatin-d7 (hydrochloride) is a deuterium-labeled version of Bestatin (hydrochloride). Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, primarily utilized in cancer research.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase
Catalog No. 50-004-50884 Supplier Medchemexpress LLC Supplier No. HYB0134AS1MG
Bestatin-d7 (hydrochloride) is a deuterium-labeled version of Bestatin (hydrochloride). Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, primarily utilized in cancer research.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase