Learn More
Research Products International Corp Bestatin Hydrochloride, 10 Milligrams
SDP
Supplier:  Research Products International Corp B121000.01
Encompass_Preferred
Competitive and specific inhibitor of leucine aminopeptidase, animo-peptidase B and triaminopeptidase.
Catalog No. 50-488-532