Learn More
Alkali Scientific Bestatin Hydrochloride, 10 Milligrams
SDP
Catalog No. 5000659861
Encompass_Preferred
Bestatin Hydrochloride is a high-purity enzyme inhibitor provided in a 10 mg quantity, used in laboratory settings for inhibiting aminopeptidases during protein analysis. This reagent is particularly useful in studies of peptide and protein metabolism where aminopeptidases play a critical role in the breakdown of peptides. Bestatin Hydrochloride is used in proteomics, cell culture studies, and enzyme inhibition research. Its application helps researchers maintain protein integrity by preventing enzymatic degradation and enhancing the precision of studies involving aminopeptidase activity. The high purity of Bestatin ensures reliability and consistency in experimental results.
Catalog No. 50-006-59861 Supplier Alkali Scientific Supplier No. BES001CP
Bestatin Hydrochloride is a high-purity enzyme inhibitor provided in a 10 mg quantity, used in laboratory settings for inhibiting aminopeptidases during protein analysis. This reagent is particularly useful in studies of peptide and protein metabolism where aminopeptidases play a critical role in the breakdown of peptides. Bestatin Hydrochloride is used in proteomics, cell culture studies, and enzyme inhibition research. Its application helps researchers maintain protein integrity by preventing enzymatic degradation and enhancing the precision of studies involving aminopeptidase activity. The high purity of Bestatin ensures reliability and consistency in experimental results.