Learn More
Apexbio Technology LLC Bestatin hydrochloride, 100mg. Cas: 65391-42-6 MFCD: MFCD00058004. Inhibitor of aminopeptidase N (APN)/CD13 and aminopeptidase B.
SDP
Catalog No. 501905168
Encompass_Preferred
Bestatin hydrochloride, also known as Ubenimex, is an inhibitor of aminopeptidase B and N (APN)/CD13 [1].Bestatin, an antibiotic of microbial origin, is a potent inhibitor of some aminopeptidases which can be administered to cultured cells, intact animals and humans with low toxicity. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5168 Supplier Apexbio Technology LLC Supplier No. A8621100
Bestatin hydrochloride, also known as Ubenimex, is an inhibitor of aminopeptidase B and N (APN)/CD13 [1].Bestatin, an antibiotic of microbial origin, is a potent inhibitor of some aminopeptidases which can be administered to cultured cells, intact animals and humans with low toxicity. Other sizes are available. Please inqury us for quote.