Learn More
Selleck Chemical LLC Bestatin hydrochloride E4916-5MG
SDP
Catalog No. 5000043391
Encompass_Preferred
Bestatin hydrochloride is an inhibitor of aminopeptidase N and enkephalinase that elevates the levels of angiotensin II and angiotensin III It also acts as a non-competitive inhibitor ( 1) of porcine DPP-IV with a Ki value of 75 M It reduces DPP-IV activity decreases cell viability and increases DNA fragmentation in vitro It has the potential to treat skin conditions like acne psoriasis and keloids
Catalog No. 50-000-43391 Supplier Selleck Chemical LLC Supplier No. E49165MG
Bestatin hydrochloride is an inhibitor of aminopeptidase N and enkephalinase that elevates the levels of angiotensin II and angiotensin III It also acts as a non-competitive inhibitor ( 1) of porcine DPP-IV with a Ki value of 75 M It reduces DPP-IV activity decreases cell viability and increases DNA fragmentation in vitro It has the potential to treat skin conditions like acne psoriasis and keloids