Learn More
Medchemexpress LLC Bestatin hydrochloride | 65391-42-6 | MFCD00058004 | 99.9% | 50 MG
SDP
Catalog No. 5000450769
Encompass_Preferred
Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, primarily used for cancer research. This compound has demonstrated a range of biological activities both in vitro and in vivo.
  • Enhances ATRA-induced differentiation
  • Inhibits ATRA-driven phosphorylation of p38 MAPK in ATRA-sensitive APL NB4 cells
  • Slowing cell cycle progression at 600 μM
  • Inhibits mitosis and multinuclearity in D. discoideum
  • Not cytotoxic at 0-600 μM
  • Reduces CD13 expression in diabetic mice
  • Inhibits MMP-9 specific gelationolytic band densities
  • Downregulates HIF-1α and VEGF expression in diabetic mice retinas
  • Increases splenocytes producing hemolytic anti-SRBC antibodies
Catalog No. 50-004-50769 Supplier Medchemexpress LLC Supplier No. HYB0134A50MG
Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, primarily used for cancer research. This compound has demonstrated a range of biological activities both in vitro and in vivo.
  • Enhances ATRA-induced differentiation
  • Inhibits ATRA-driven phosphorylation of p38 MAPK in ATRA-sensitive APL NB4 cells
  • Slowing cell cycle progression at 600 μM
  • Inhibits mitosis and multinuclearity in D. discoideum
  • Not cytotoxic at 0-600 μM
  • Reduces CD13 expression in diabetic mice
  • Inhibits MMP-9 specific gelationolytic band densities
  • Downregulates HIF-1α and VEGF expression in diabetic mice retinas
  • Increases splenocytes producing hemolytic anti-SRBC antibodies