Learn More
Abcam Bestatin hydrochloride, aminopeptidase inhibitor, 1MG
Catalog No. 502706255
Encompass_Preferred
MW 344.8 Da, Purity >98%. Potent and specific aminopeptidase inhibitor. Antibiotic. Inhibits 12 aminopeptidases including aminopeptidase B, (Ki = 1 μM), leucyl aminopeptidase (Ki = 1 nM) and cytosol nonspecific dipeptidase (Ki = 4 nM). Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6255 Supplier Abcam Supplier No. AB1424051MG
MW 344.8 Da, Purity >98%. Potent and specific aminopeptidase inhibitor. Antibiotic. Inhibits 12 aminopeptidases including aminopeptidase B, (Ki = 1 μM), leucyl aminopeptidase (Ki = 1 nM) and cytosol nonspecific dipeptidase (Ki = 4 nM). Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement