Learn More
Medchemexpress LLC Bestatin hydrochloride (Ubenimex hydrochloride) | 65391-42-6 | MFCD00058004 | 99.9% | 100 MG
SDP
Catalog No. 5000450943
Encompass_Preferred
Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research. Bestatin hydrochloride is also known to have several biological activities, including:
  • Inhibitor of CD13 (aminopeptidase N)/APN and leukotriene A4 hydrolase
  • Enhances ATRA-induced differentiation in ATRA-sensitive APL NB4 cells
  • Inhibits ATRA-driven phosphorylation of p38 MAPK
  • Slows cell cycle progression in D. discoideum
  • Inhibits mitosis and inherent multinuclearity in D. discoideum
  • Reduces CD13 expression in diabetic mice
  • Inhibits MMP-9 specific gelationolytic band densities in diabetic mice
  • Inhibits VEGF and heparanase expression in diabetic mice
  • Increases splenocytes producing hemolytic anti-SRBC antibodies (at 0.1 mg/kg)
Catalog No. 50-004-50943 Supplier Medchemexpress LLC Supplier No. HYB0134A100MG
Bestatin hydrochloride is an inhibitor of CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase, used for cancer research. Bestatin hydrochloride is also known to have several biological activities, including:
  • Inhibitor of CD13 (aminopeptidase N)/APN and leukotriene A4 hydrolase
  • Enhances ATRA-induced differentiation in ATRA-sensitive APL NB4 cells
  • Inhibits ATRA-driven phosphorylation of p38 MAPK
  • Slows cell cycle progression in D. discoideum
  • Inhibits mitosis and inherent multinuclearity in D. discoideum
  • Reduces CD13 expression in diabetic mice
  • Inhibits MMP-9 specific gelationolytic band densities in diabetic mice
  • Inhibits VEGF and heparanase expression in diabetic mice
  • Increases splenocytes producing hemolytic anti-SRBC antibodies (at 0.1 mg/kg)