Learn More
Expression Systems BestBac 2.0 Δ v-cath/chiA Baculovirus Cotransfection Kit
SDP
Catalog No. NC9909291
Encompass_Preferred
Expression Systems popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors.
Unmatched productivity and stability for BEVS
Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/mL of recombinant baculovirus in P1 stock
Generates P1 stock as quickly as Bac-to-Bac and other systems
Each BestBac vial contains enough material for a minimum of five cotransfections
Suitable for other expression systems including BacMam™
Contains everything you need to perform cotransfections with your transfer vector. Included in the kit are:
• BestBac DNA
• pVL1393-lacZ control vector
• Transfection medium
Expres2 TR transfection reagent
To download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. NC9909291 Supplier Expression Systems Supplier No. 91200
Expression Systems popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors.
Unmatched productivity and stability for BEVS
Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/mL of recombinant baculovirus in P1 stock
Generates P1 stock as quickly as Bac-to-Bac and other systems
Each BestBac vial contains enough material for a minimum of five cotransfections
Suitable for other expression systems including BacMam™
Contains everything you need to perform cotransfections with your transfer vector. Included in the kit are:
• BestBac DNA
• pVL1393-lacZ control vector
• Transfection medium
Expres2 TR transfection reagent
To download Instructions for Use and/or COA please visit: www.expressionsystems.com