Learn More
Expression Systems BestBac 2.0 V-Cath/ChiA Deleted Baculovirus DNA
SDP
Catalog No. NC9840159
Encompass_Preferred
Expression Systems popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors.
Unmatched productivity and stability for BEVS
Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/mL of recombinant baculovirus in P1 stock
Generates P1 stock as quickly as Bac-to-Bac and other systems
Each BestBac vial contains enough material for a minimum of five cotransfections
Suitable for other expression systems including BacMam™
BestBac 2.0 is a deletion mutant of BestBac 1.0. The v-cath/chiA deletion removes a potentially detrimental protease and may reduce competition for resources for protein synthesis during late gene expression. The v-cath/chiA deletion can improve the integrity of your expressed protein resulting in a significant increase of functional protein yield.
Catalog No. NC9840159 Supplier Expression Systems Supplier No. 91002
Expression Systems popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors.
Unmatched productivity and stability for BEVS
Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/mL of recombinant baculovirus in P1 stock
Generates P1 stock as quickly as Bac-to-Bac and other systems
Each BestBac vial contains enough material for a minimum of five cotransfections
Suitable for other expression systems including BacMam™
BestBac 2.0 is a deletion mutant of BestBac 1.0. The v-cath/chiA deletion removes a potentially detrimental protease and may reduce competition for resources for protein synthesis during late gene expression. The v-cath/chiA deletion can improve the integrity of your expressed protein resulting in a significant increase of functional protein yield.