Learn More
Andwin Scientific BESTRACK 96-PLACE REVERSIBLE
SDP
Catalog No. 50820010
Encompass_Preferred
This tube rack provides secure storage for up to 96 1.5 and 2.0 mL tubes. This product features alphanumeric sample identification that permits easy organization. It includes a lid that has a snug fit and has a tall height to give space for screw-cap microcentrifuge tubes. This product is sold in a pack of 5.
Catalog No. 50-820-010 Supplier Andwin Scientific Supplier No. 23965048
This tube rack provides secure storage for up to 96 1.5 and 2.0 mL tubes. This product features alphanumeric sample identification that permits easy organization. It includes a lid that has a snug fit and has a tall height to give space for screw-cap microcentrifuge tubes. This product is sold in a pack of 5.