Learn More
Andwin Scientific Green Polypropylene 96-place tube BestRack, 8 7/16"" L x 4 5/8"" W x 2"" H, 5/pack
SDP
Catalog No. 50820000
Encompass_Preferred
Holds 1.5 and 2.0 ml tubes on one side or 0.5 ml tubes on the other.Alphanumeric sample identification.Withstands freezing (-90C) or autoclaving.BestRack reversible rack holds 96 samples in an 8 x 12 array. Optimum spacing provides easy access to each tube, even when all 96 places are full. The transparent lid has a snug fit and is tall enough to store most screw-cap microcentrifuge tubes. Mixed neon pack includes blue, green, orange, pink, and yellow racks with natural lids. Measures 8 7/16" L x 4 3/8" W x 2" H with lid and has frosted labeling areas on four sides. Polypropylene.
Catalog No. 50-820-000 Supplier Andwin Scientific Supplier No. 23965002
Holds 1.5 and 2.0 ml tubes on one side or 0.5 ml tubes on the other.Alphanumeric sample identification.Withstands freezing (-90C) or autoclaving.BestRack reversible rack holds 96 samples in an 8 x 12 array. Optimum spacing provides easy access to each tube, even when all 96 places are full. The transparent lid has a snug fit and is tall enough to store most screw-cap microcentrifuge tubes. Mixed neon pack includes blue, green, orange, pink, and yellow racks with natural lids. Measures 8 7/16" L x 4 3/8" W x 2" H with lid and has frosted labeling areas on four sides. Polypropylene.