Learn More
JASCO INC JASCO BeStSel CFR Program, Advanced Secondary Structure Estimation, Compatible With JASCO CD Spectrometers
SDP
Catalog No. NC3716704
Encompass
The JASCO BeStSel CFR Program is an advanced application software designed for accurate secondary structure estimation of proteins through Circular Dichroism (CD) spectroscopy. It offers robust control and analysis for JASCO CD spectrometers, leveraging the BeStSel algorithm to enhance precision by considering β-strand orientation and the twist of antiparallel β-sheets. With GxP compliance and batch processing capabilities, it facilitates objective evaluation and comprehensive biomolecular structural analysis.
Catalog No. NC3716704 Supplier JASCO INC Supplier No. 4880J141A
Add to Cart
Add to Cart
Request a Quote
The JASCO BeStSel CFR Program is an advanced application software designed for accurate secondary structure estimation of proteins through Circular Dichroism (CD) spectroscopy. It offers robust control and analysis for JASCO CD spectrometers, leveraging the BeStSel algorithm to enhance precision by considering β-strand orientation and the twist of antiparallel β-sheets. With GxP compliance and batch processing capabilities, it facilitates objective evaluation and comprehensive biomolecular structural analysis.