Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bet-bay 002 | 1588521-78-1 | 99.8% | 403.86 g/mol | C22H18ClN5O | 100MG
SDP

Catalog No. 5000236091
Encompass_Preferred

BET-BAY 002 is a small-molecule BET bromodomain inhibitor intended for research use. It demonstrates activity in multiple myeloma models and is provided as a high-purity solid for in vitro and preclinical studies. The compound is soluble in DMSO and is offered in multiple pack sizes to support assay development and screening.

  • Potent BET bromodomain inhibitor with reported efficacy in multiple myeloma models.
  • High purity (99.8%) suitable for biological assays.
  • Soluble in DMSO for convenient reconstitution.
  • Available in multiple quantities including a 100 mg package.
  • Molecular weight 403.86 g/mol and formula C22H18ClN5O.

Catalog No. 50-002-36091 Supplier Medchemexpress LLC Supplier No. HY12421100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BET-BAY 002 is a small-molecule BET bromodomain inhibitor intended for research use. It demonstrates activity in multiple myeloma models and is provided as a high-purity solid for in vitro and preclinical studies. The compound is soluble in DMSO and is offered in multiple pack sizes to support assay development and screening.

  • Potent BET bromodomain inhibitor with reported efficacy in multiple myeloma models.
  • High purity (99.8%) suitable for biological assays.
  • Soluble in DMSO for convenient reconstitution.
  • Available in multiple quantities including a 100 mg package.
  • Molecular weight 403.86 g/mol and formula C22H18ClN5O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.