Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bet-bay 002 | 1588521-78-1 | 99.8% | 403.86 g/mol | C22H18ClN5O | 10MM 1ML
SDP

Catalog No. 5000239049
Encompass_Preferred

BET-BAY 002 is a small-molecule inhibitor of BET family bromodomains used for epigenetic and oncology research. It has demonstrated efficacy in multiple myeloma models and is supplied in solution and solid forms suitable for biochemical and cellular assays.

  • Supplied as a 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (99.8%).
  • Molecular weight 403.86 g/mol and formula C22H18ClN5O.
  • Soluble in DMSO for straightforward stock preparation.
  • Intended for research use in BET bromodomain inhibition and related assays.

Catalog No. 50-002-39049 Supplier Medchemexpress LLC Supplier No. HY1242110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BET-BAY 002 is a small-molecule inhibitor of BET family bromodomains used for epigenetic and oncology research. It has demonstrated efficacy in multiple myeloma models and is supplied in solution and solid forms suitable for biochemical and cellular assays.

  • Supplied as a 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (99.8%).
  • Molecular weight 403.86 g/mol and formula C22H18ClN5O.
  • Soluble in DMSO for straightforward stock preparation.
  • Intended for research use in BET bromodomain inhibition and related assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.