Learn More
Medchemexpress LLC BET bromodomain inhibitor 1 | 2411226-02-1 | 99.7% | 100 MG
SDP
Catalog No. 5000339886
Encompass_Preferred
BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor with an IC50 of 2.6 nM for BRD4. It binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities (Kd values of 1.3 nM, 1.0 nM, 3.0 nM, 1.6 nM, 2.1 nM, respectively). This inhibitor demonstrates anti-cancer activity.
  • Orally active, selective BET bromodomain inhibitor
  • High affinity binding to BRD proteins (BRD2(2), BRD3(2), BRD4(1), BRD4(2), BRDT(2))
  • Demonstrates anti-cancer activity
  • Excellent selectivity for BET bromodomain family
  • Does not inhibit five cytochrome P450 enzymes
  • Inhibits growth of acute myeloid leukemia and multiple myeloma cell lines
  • Exhibits strong antitumor activities in vivo
  • Induces G1-phase cell cycle arrest
  • Dose-dependent inhibition of c-Myc and upregulation of p21
Catalog No. 50-003-39886 Supplier Medchemexpress LLC Supplier No. HY131061100MG
BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor with an IC50 of 2.6 nM for BRD4. It binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities (Kd values of 1.3 nM, 1.0 nM, 3.0 nM, 1.6 nM, 2.1 nM, respectively). This inhibitor demonstrates anti-cancer activity.
  • Orally active, selective BET bromodomain inhibitor
  • High affinity binding to BRD proteins (BRD2(2), BRD3(2), BRD4(1), BRD4(2), BRDT(2))
  • Demonstrates anti-cancer activity
  • Excellent selectivity for BET bromodomain family
  • Does not inhibit five cytochrome P450 enzymes
  • Inhibits growth of acute myeloid leukemia and multiple myeloma cell lines
  • Exhibits strong antitumor activities in vivo
  • Induces G1-phase cell cycle arrest
  • Dose-dependent inhibition of c-Myc and upregulation of p21