Learn More
Medchemexpress LLC BET bromodomain inhibitor 1 | 2411226-02-1 | 99.7% | 50 MG
SDP
Catalog No. 5000338568
Encompass_Preferred
BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor. It demonstrates high affinity binding to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) and exhibits potent anti-cancer activity.
  • Orally active, selective BET bromodomain inhibitor
  • IC50 of 2.6 nM for BRD4
  • Binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities
  • Exhibits potent anti-cancer activity
  • Induces G1-phase cell cycle arrest
  • Inhibits c-Myc expression and upregulates p21 levels
  • Reduces c-Myc, BCL-2, and CDK6 expressions
  • Highly selective for BET bromodomain family (~1500-fold over EP300)
  • Strong antitumor activities in MV4-11 mouse xenograft model
Catalog No. 50-003-38568 Supplier Medchemexpress LLC Supplier No. HY13106150MG
BET bromodomain inhibitor 1 is an orally active, selective bromodomain and extra-terminal (BET) bromodomain inhibitor. It demonstrates high affinity binding to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) and exhibits potent anti-cancer activity.
  • Orally active, selective BET bromodomain inhibitor
  • IC50 of 2.6 nM for BRD4
  • Binds to BRD2(2), BRD3(2), BRD4(1), BRD4(2), and BRDT(2) with high affinities
  • Exhibits potent anti-cancer activity
  • Induces G1-phase cell cycle arrest
  • Inhibits c-Myc expression and upregulates p21 levels
  • Reduces c-Myc, BCL-2, and CDK6 expressions
  • Highly selective for BET bromodomain family (~1500-fold over EP300)
  • Strong antitumor activities in MV4-11 mouse xenograft model