Learn More
Medchemexpress LLC Bet bromodomain inhibitor 1 | 2411226-02-1 | 99.9% | C22H19F2N3O4S | 10MG
SDP
Catalog No. 5000261739
Encompass_Preferred
BET bromodomain inhibitor 1 is an orally active, selective BET family bromodomain inhibitor used in epigenetic and anticancer research. It binds BRD4 with low-nanomolar potency and shows antiproliferative activity in multiple human cancer cell lines.
  • Potent BRD4 inhibition (IC50 = 2.6 nM).
  • Low-nanomolar binding to BET family members (Kd values ~1-3 nM).
  • High purity (≈99.9%) suitable for research applications.
  • Defined molecular formula C22H19F2N3O4S and molecular weight 459.47 g/mol.
  • Demonstrated antiproliferative activity in cancer cell lines.
  • Available in small research pack sizes and as a DMSO stock solution.
Catalog No. 50-002-61739 Supplier Medchemexpress LLC Supplier No. HY13106110MG
BET bromodomain inhibitor 1 is an orally active, selective BET family bromodomain inhibitor used in epigenetic and anticancer research. It binds BRD4 with low-nanomolar potency and shows antiproliferative activity in multiple human cancer cell lines.
  • Potent BRD4 inhibition (IC50 = 2.6 nM).
  • Low-nanomolar binding to BET family members (Kd values ~1-3 nM).
  • High purity (≈99.9%) suitable for research applications.
  • Defined molecular formula C22H19F2N3O4S and molecular weight 459.47 g/mol.
  • Demonstrated antiproliferative activity in cancer cell lines.
  • Available in small research pack sizes and as a DMSO stock solution.