Learn More
Medchemexpress LLC BET bromodomain inhibitor | 1505453-59-7 | MFCD28144506 | 99.5% | 445.90 | C24H20ClN5O2 | 1 ML
SDP
Catalog No. 5000247521
Encompass_Preferred
BET bromodomain inhibitor is an orally active, selective BET-family bromodomain inhibitor used in cancer research to inhibit tumor cell proliferation, induce cell cycle arrest, and promote apoptosis. It is supplied as a solid and as a 10 mM solution in DMSO (1 mL). The compound has a molecular formula of C24H20ClN5O2 and a molecular weight of 445.90.
  • Orally active, selective BET bromodomain inhibitor
  • Available as a solid or 10 mM solution in DMSO (1 mL)
  • Used in cell-based assays to study proliferation, cell cycle, and apoptosis
  • High chemical purity suitable for research applications
  • Store powder and solutions as recommended to preserve stability
Catalog No. 50-002-47521 Supplier Medchemexpress LLC Supplier No. HY10303610MM 1ML
BET bromodomain inhibitor is an orally active, selective BET-family bromodomain inhibitor used in cancer research to inhibit tumor cell proliferation, induce cell cycle arrest, and promote apoptosis. It is supplied as a solid and as a 10 mM solution in DMSO (1 mL). The compound has a molecular formula of C24H20ClN5O2 and a molecular weight of 445.90.
  • Orally active, selective BET bromodomain inhibitor
  • Available as a solid or 10 mM solution in DMSO (1 mL)
  • Used in cell-based assays to study proliferation, cell cycle, and apoptosis
  • High chemical purity suitable for research applications
  • Store powder and solutions as recommended to preserve stability