Learn More
Medchemexpress LLC Bet bromodomain inhibitor | 1505453-59-7 | MFCD28144506 | 99.5% | 445.90 | C24H20ClN5O2 | 5 MG
SDP
Catalog No. 5000246288
Encompass_Preferred
BET bromodomain inhibitor (CAS 1505453-59-7) is a high-purity research compound that targets BET family bromodomains, with reported activity against BRD4 (IC50 = 14 nM). Supplied as a small solid pack for laboratory use, it is intended for in vitro and preclinical studies of epigenetic regulation and tumor cell biology.
  • High purity (99.53%).
  • Reported BRD4 potency: IC50 = 14 nM.
  • Molecular formula C24H20ClN5O2; molecular weight 445.90.
  • Supplied as a 5 mg solid suitable for assay development.
  • Useful for epigenetic and tumor research applications.
Catalog No. 50-002-46288 Supplier Medchemexpress LLC Supplier No. HY1030365MG
BET bromodomain inhibitor (CAS 1505453-59-7) is a high-purity research compound that targets BET family bromodomains, with reported activity against BRD4 (IC50 = 14 nM). Supplied as a small solid pack for laboratory use, it is intended for in vitro and preclinical studies of epigenetic regulation and tumor cell biology.
  • High purity (99.53%).
  • Reported BRD4 potency: IC50 = 14 nM.
  • Molecular formula C24H20ClN5O2; molecular weight 445.90.
  • Supplied as a 5 mg solid suitable for assay development.
  • Useful for epigenetic and tumor research applications.