Learn More
Medchemexpress LLC Bet bromodomain inhibitor | 1505453-59-7 | MFCD28144506 | 99.5% | 445.90 g/mol | C24H20ClN5O2 | 100 MG
SDP
Catalog No. 5000246179
Encompass_Preferred
BET bromodomain inhibitor is a small-molecule research compound that selectively inhibits BET family bromodomains, including BRD4, with reported biochemical potency (BRD4(1) IC50 = 14 nM). It is supplied as a solid (white to off-white) with high purity for preclinical laboratory use.
  • Potent BRD4 inhibitor (IC50 = 14 nM).
  • High purity (~99.5%).
  • Solid, white to off-white powder.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities, including 100 MG packaging.
Catalog No. 50-002-46179 Supplier Medchemexpress LLC Supplier No. HY103036100MG
BET bromodomain inhibitor is a small-molecule research compound that selectively inhibits BET family bromodomains, including BRD4, with reported biochemical potency (BRD4(1) IC50 = 14 nM). It is supplied as a solid (white to off-white) with high purity for preclinical laboratory use.
  • Potent BRD4 inhibitor (IC50 = 14 nM).
  • High purity (~99.5%).
  • Solid, white to off-white powder.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research quantities, including 100 MG packaging.